missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL4I1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5106825
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody detects endogenous levels of total IL4I1.
This gene encodes a protein with limited similarity to L-amino acid oxidase which contains the conserved amino acids thought to be involved in catalysis and binding of flavin adenine dinucleotide (FAD) cofactor. The expression of this gene can be induced by interleukin 4 in B cells, however, expression of transcripts containing the first two exons of the upstream gene is found in other cell types. Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification
| IL4I1 | |
| Polyclonal | |
| Unconjugated | |
| IL4I1 | |
| FIG1; Fig-1 protein; Fig1-ps; hFIG1; IL4I1; IL4I1_2; IL4-induced protein 1; interleukin 4 induced 1; interleukin four induced 1; interleukin-4-induced protein 1; interleukin-four induced gene 1; interleukin-four induced gene-1 protein; interleukin-four induced protein-1; LAAO; L-amino acid oxidase; L-amino-acid oxidase; LAO; LOC100360621; mFIG1; Protein Fig-1; rCG53598-like; UNQ636/PRO1265 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 259307 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96RQ9 | |
| IL4I1 | |
| A synthesized peptide derived from human IL4I1(Accession Q96RQ9), corresponding to amino acid residues M1-W51. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu