missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ING3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA540702
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Peptide sequence: MDQLEQRVSE FFMNAKKNKP EWREEQMASI KKDYYKALED ADEKVQLANQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
The protein encoded by this gene is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This gene can activate p53 trans-activated promoters, including promoters of p21/waf1 and bax. Overexpression of this gene has been shown to inhibit cell growth and induce apoptosis. Allelic loss and reduced expression of this gene were detected in head and neck cancers. Two alternatively spliced transcript variants encoding different isoforms have been observed.
Spécification
| ING3 | |
| Polyclonal | |
| Unconjugated | |
| ING3 | |
| 1300013A07Rik; Eaf 4; Eaf4; FLJ20089; HSPC301; ING 2; ING 3; ING2; ING3; ing3 protein; inhibitor of growth family member 3; inhibitor of growth family, member 3; inhibitor of growth protein 3; inhibitor of growth protein 3-like protein; MEAF4; p47 p47 ING3; p47 regulator protein; p47ING3; p47ING3 protein; zgc:56327 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 54556 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NXR8 | |
| ING3 | |
| Synthetic peptide directed towards the N-terminal of human ING3 (aa 36-85). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu