missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA554191
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: KEKRKLLMQN QSSTNHPGAS IALSRPSLNK DFRDHAEQQH IAAQQKAALQ HAHAHSSGYF ITQDSAFGNL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
Spécification
| INIP | |
| Polyclonal | |
| Unconjugated | |
| INIP | |
| 1110054O05Rik; 2610312O17Rik; AA399876; C9orf80; HSPC043; HSPC291; hSSB-interacting protein 1; hSSBIP1; Inip; INTS3 and NABP interacting protein; INTS3- and NABP-interacting protein; minute INTS3/hSSB-associated element; MISE; RGD1307983; Sensor of single-strand DNA complex subunit C; sensor of ssDNA subunit C; Single-stranded DNA-binding protein-interacting protein 1; SOSS complex subunit C; SOSSC; SOSS-C; SSB-interacting protein 1; Ssbip1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 58493 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NRY2 | |
| INIP | |
| Recombinant protein corresponding to Human INIP. Recombinant protein control fragment (Product #RP-92999). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu