missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Kit de marquage des anticorps avec nanoparticules en or InnovaGOLD, 40 nm, dia. ext. 20
High efficiency antibody binding for maximum sensitivity
Marque: Novus Biologicals™ 220-0010
Informations supplémentaires : Poids : 0.00970kg
Alert:
For Research Use Only
Description
- Uniform Shape
- Low co-efficient of variation in terms of size
- High efficiency antibody binding for maximum sensitivity
- Available at high concentrations to simplify assay development
Spécification
Store at 4°C. Do not freeze. | |
10 mL | |
40nm Colloidal Gold Nanoparticles 20 OD | |
Antibody Labeling Kit |