Learn More
Integrin alpha 4 beta 7/LPAM-1 Antibody (CL11293), Novus Biologicals™
Mouse Monoclonal Antibody
329.00€ - 600.00€
Spécification
| Antigène | Integrin alpha 4 beta 7/LPAM-1 |
|---|---|
| Clone | CL11293 |
| Dilution | Western Blot 1 ug/ml |
| Applications | Western Blot |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
18622585
|
Novus Biologicals
NBP3-21189-100ul |
100 μg |
600.00€
100µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
18654664
|
Novus Biologicals
NBP3-21189-25ul |
25 μg |
329.00€
25µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Integrin alpha 4 beta 7/LPAM-1 Monoclonal antibody specifically detects Integrin alpha 4 beta 7/LPAM-1 in Human samples. It is validated for Western BlotSpécification
| Integrin alpha 4 beta 7/LPAM-1 | |
| Western Blot 1 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
| This antibody was developed against a Synthetic Peptide of P13612. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL11293 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| PBS, pH 7.2, 40% glycerol | |
| 3676 | |
| IgG1 | |
| Protein A purified |