Learn More
Integrin alpha 4/CD49d Antibody (CL11294) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP3-07999PE
Code nomenclature Nacres: NA.46
Code produit. 30291845
  • 737.00€ / 0.10mL
Description
Integrin alpha 4/CD49d Monoclonal antibody specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Integrin alpha 4/CD49d
Monoclonal
PE
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
This antibody was developed using a synthetic peptide derived from P13612, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Microautophagy, Neuronal Cell Markers, Phospho Specific
3676
Store at 4⪚C in the dark.
IgG2b
Western Blot
CL11294
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified