Learn More
Integrin alpha V beta 6 Antibody (STX-100), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals (Bio-Techne) NBP3-28326CL1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Integrin alpha V beta 6 Monoclonal antibody specifically detects Integrin alpha V beta 6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| Integrin alpha V beta 6 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD51, integrin subunit alpha V, ITGAV, MSK8, VNRA, VTNR | |
| Integrin aVb6 (ITGAV & ITGB6) | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Signaling, Stem Cells | |
| 3685 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| STX-100 | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |