missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Interferon beta Monoclonal Antibody (C2), Invitrogen™

Mouse Monoclonal Antibody
180.00€ - 453.00€
Spécification
| Antigène | Interferon beta |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
17674206
|
Invitrogen™
MA543753 |
20 μL |
180.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
17664206
![]() |
Invitrogen™
MA543752 |
100 μL |
453.00€
100µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.Spécification
| Interferon beta | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1 | |
| Ifnb1 | |
| IgG2a κ | |
| Protein A/G | |
| Antibody |
| C1 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01574 | |
| 3456 | |
| Met22-Asn187. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Ifnb1 |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit