missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Involucrin Monoclonal Antibody (SY8), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA190573
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA1-90573 detects Involucrin from non-human primate, porcine, and human samples.
Involucrin, a component of the keratinocyte crosslinked envelope, is found in the cytoplasm and crosslinked to membrane proteins by transglutaminase. Involucrin is expressed in a range of stratified squamous epithelia, including the cornea. In normal epidermis, it is first expressed in the upper spinous layers, and in keratinocyte cultures it is expressed by all cells that have left the basal layer. Involucrin expression is abnormal in squamous cell carcinomas and premalignant lesions and is reduced in severe dysplasias of the larynx and cervix. Its gene is mapped to 1q21, among calpactin I light chain, trichohyalin, profillaggrin, loricrin, and calcyclin.
Spécification
| Involucrin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P07476, P18174, P18175 | |
| IVL | |
| Pure involucrin from human keratinocytes. | |
| 100 μg | |
| Primary | |
| Avian, Canine, Human, Non-human Primate, Pig | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| SY8 | |
| Unconjugated | |
| IVL | |
| 1110019C06Rik; involucrin; IVL | |
| Mouse | |
| Protein G | |
| RUO | |
| 101125010, 102152041, 3713, 407242 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu