Learn More
IRF4 Antibody (CL14615) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-26863JF549
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IRF4 Monoclonal antibody specifically detects IRF4 in Human samples. It is validated for Immunohistochemistry,Western Blot
Spécification
| IRF4 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5 | |
| This antibody was developed using a recombinant protein derived from Q15306, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 3662 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| CL14615 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |