Learn More
Irisin/FNDC5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Marque: Novus Biologicals NBP2-14024
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Irisin/FNDC5 Polyclonal specifically detects Irisin/FNDC5 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Spécification
| Irisin/FNDC5 | |
| Western Blot, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50, Immunofluorescence Reported in scientific literature (PMID:31614634). | |
| FNDC5 | |
| Affinity Purified |
| Immunohistochemistry | |
| fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FRCP2Fibronectin type III repeat-containing protein 2 | |
| Rabbit | |
| 0.1 mL |