missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Iscalimab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA542187
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Iscalimab (CFZ533) is a new, fully humanized monoclonal antibody that targets cluster of differentiation 40 (CD40), which can regulate the CD40-CD154 costimulatory pathway and prevent CD40 pathway signal transduction and CD40+ cell type activation. It is expected Become a new treatment for solid organs and related autoimmune diseases.
Spécification
| Iscalimab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CFZ-533; NVP-CFZ533; OM11-62MF | |
| Human CD40/TNFRSF5. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 1.24 mg/mL | |
| PBS with no preservative; pH 7.5 | |
| Human | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu