missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KANSL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA585459
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Keep as concentrated solution. Predicted reactivity: Mouse (89%). Positive Control: THP-1 whole cell extract, THP-1 nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a nuclear protein that is a subunit of two protein complexes involved with histone acetylation, the MLL1 complex and the NSL1 complex. The corresponding protein in Drosophila interacts with K(lysine) acetyltransferase 8, which is also a subunit of both the MLL1 and NSL1 complexes.
Spécification
| KANSL1 | |
| Polyclonal | |
| Unconjugated | |
| KANSL1 | |
| 1700081L11Rik; 9430041J06Rik; CENP-36; centromere protein 36; centromere protein 36 antibody; DKFZp686P06109 antibody; DKFZp727C091 antibody; hMSL1v1; Kansl1; KAT8 regulatory NSL complex subunit 1; KAT8 regulatory NSL complex subunit 1 antibody; KDVS; KIAA1267; KIAA1267 antibody; male-specific lethal 1 homolog; MGC102843 antibody; mKIAA1267; MLL1/MLL complex subunit KANSL1; MLL1/MLL complex subunit male-specific lethal 1 homolog antibody; MSL1 homolog 1; MSL1V1; MSL1v1 antibody; non-specific lethal 1 homolog; NSL complex protein NSL1; NSL1; RGD1311429 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 284058 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.82 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q7Z3B3 | |
| KANSL1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human KIAA1267. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu