missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir3.3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-57515-25ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Kir3.3 Polyclonal specifically detects Kir3.3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| Kir3.3 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| G protein-activated inward rectifier potassium channel 3, G protein-coupled inward rectifier potassium channel, GIRK-3, GIRK3KIR3.3, Inward rectifier K(+) channel Kir3.3, inwardly rectifier K+ channel KIR3.3, Kir3.3, Potassium channel, inwardly rectifying subfamily J member 9, potassium inwardly-rectifying channel, subfamily J, member 9 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3765 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| KCNJ9 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RLDAHLYWSIPSRLDEKVEEE | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu