missing translation for 'onlineSavingsMsg'
Learn More

KSHV ORF57 Antibody, Novus Biologicals™

Code produit 18063907 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
50 μg
Unit Size:
100µg
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18063907 100 μg 100µg
18067146 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18063907 Supplier Novus Biologicals Supplier No. NBP295260

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KSHV ORF57 Polyclonal specifically detects KSHV ORF57 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigène KSHV ORF57
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1.55 mg/ml
Conjugué Unconjugated
Dilution Western Blot 1:5000-1:8000, ELISA 1:100000-170000, Immunohistochemistry
Formule 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Numéro d’ordre du gène Q2HR75
Alias de gène herpesvirus core gene UL54 family, ORF57
Symboles de gène(s) ORF57
Espèces hôtes Rabbit
Immunogène This affinity purified KSHV ORF57 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a region near the C-terminal of human KSHV ORF57 protein. (Uniprot: Q2HR75)
Poids moléculaire de l’antigène 51.01 kDa
Méthode de purification Immunogen affinity purified
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 4961525
Espèces cibles Virus
Contenu et stockage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.