Learn More
Ku80 Monoclonal Antibody (111), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA512933
Description
MA5-12933 targets Ku (p80) in FACS, ICC/IF, IP, and WB applications and shows reactivity with Human, Mouse and Non-human primate samples. This antibody does not cross-react with mouse in immunofluorescent applications, but does react with mouse in FACS applications. The MA5-12933 immunogen is human B cell nuclei from plasmacytoid 2p68 cells.
The protein encoded by this gene is the 80-kilodalton subunit of the Ku heterodimer protein which is also known as ATP-dependant DNA helicase II or DNA repair protein XRCC5. Ku is the DNA-binding component of the DNA-dependent protein kinase, and it functions together with the DNA ligase IV-XRCC4 complex in the repair of DNA double-strand break by non-homologous end joining and the completion of V(D)J recombination events. This gene functionally complements Chinese hamster xrs-6, a mutant defective in DNA double-strand break repair and in ability to undergo V(D)J recombination. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity.
Spécification
| Ku80 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P13010, P27641 | |
| XRCC5 | |
| Human B cell nuclei from plasmacytoid 2p68 cells | |
| 500 μL | |
| Primary | |
| Human, Mouse, Non-human Primate | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 111 | |
| Unconjugated | |
| XRCC5 | |
| 86 kDa subunit of Ku antigen; AI314015; ATP-dependent DNA helicase 2 subunit 2; ATP-dependent DNA helicase II 80 kDa subunit; CTC box-binding factor 85 kDa subunit; CTC85; CTCBF; DNA repair protein XRCC5; FLJ39089; G22p2; HGNC:12833; KARP1; KARP-1; Ku autoantigen; ku autoantigen protein p86 homolog; Ku autoantigen, 80kD); Ku autoantigen, 80kDa; Ku p80; Ku80; Ku-80; Ku86; Ku-86; Ku86 autoantigen related protein 1; KUB2; Kup80; Lupu; lupus; lupus Ku autoantigen protein p86; NFIV; nuclear factor IV; OTTHUMP00000164061; OTTHUMP00000206791; Thyroid-lupus autoantigen; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 5; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining; Ku autoantigen, 80kDa); X-ray repair cross complementation (double-strand-break rejoining; X-ray repair cross complementation (double-strand-break | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 22596, 7520 | |
| 4° C | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.