Learn More
Amyloid Precursor Protein (APP) Rabbit Polyclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.
95.55€ - 645.00€
Spécification
| Antigène | Amyloid Precursor Protein (APP) |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Espèces hôtes | Rabbit |
| Code produit | Marque | Quantité | Forme | Prix | Quantité et disponibilité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Forme | Prix | Quantité et disponibilité | ||||
|
12663337
|
Epredia
RB-9023-P0 |
100 μL | Concentrated | ||||||
|
12633337
|
Epredia
RB-9023-P1 |
500 μL | Concentrated | ||||||
|
12623337
|
Epredia
RB-9023-P |
1 mL | Concentrated | ||||||
|
12673337
|
Epredia
RB-9023-R7 |
7 mL | Prediluted | ||||||
Description
Recognizes APP (b-APP770), MW 110kDa. Predicted to recognize Β-APP695 and Β-APP751. Amyloid precursor protein (APP) and APP-like proteins are transmembrane glycoproteins with a similar modular domain structure.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: N-terminal
Immunogen: A synthetic peptide corresponding to N-terminal of human APP
Molecular Weight: 110kDa
Positive Control: Brain
Cellular Localization: Cytoplasmic and extracellular
This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Spécification
| Amyloid Precursor Protein (APP) | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| IgG |
| Western Blot | |
| Unconjugated | |
| IVD | |
| A synthetic peptide corresponding to N-terminal of human APP | |
| Primary |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.