Learn More
Epredia™ Lab Vision™ CA19-9/Sialyl Lewisa (GI Tumor Marker) Mouse Monoclonal Antibody with BSA and azide
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CA19-9/Sialyl Lewisa (GI Tumor Marker), Mouse Monoclonal Antibody.
227.00€
Spécification
| Antigène | CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) |
|---|---|
| Clone | SPM110 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugué | Unconjugated |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
12662587
|
Epredia™
MS-9085-R7 |
7 mL | |||||||
Description
Mucin glucoprotein is a sialyl Lewisa structure which is synthesized from type 1 blood group precursor chains and is present in individuals expressing the Lewisa and/or Lewisb blood group antigens. In normal tissues, sialyl Lewisa antigen is present in ductal epithelium of the breast, kidney, salivary gland, and sweat glands. Its expression is greatly enhanced in serum as well as in the majority of tumor cells in gastrointestinal (GI) carcinomas.Host Species: Mouse
Clone: SPM110
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Immunogen: Purified human CA19-9
Molecular Weight: >400kDa
Positive Control: Colon CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Spécification
| CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Oncogenesis | |
| Purified human CA19-9 | |
| Primary |
| SPM110 | |
| Monoclonal | |
| Prediluted | |
| IVD | |
| Human | |
| IgG1 κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.