Learn More
Invitrogen™ Laminin alpha-4 Monoclonal Antibody (CL3183)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA524650
Description
Immunogen sequence: ENLLNQAREL QAKAESSSDE AVADTSRRVG GALARKSALK TRLSDAVKQL QAAERGDAQQ RLGQSRLITE EANRTTMEVQ QATAPMANNL TNWSQNLQHF DSSAYNTAVN SARDAVRNLT EVVPQLLDQL RTVEQKRPAS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%. Binds to an epitope located within the peptide sequence MANNLTNWSQNLQHF as determined by overlapping synthetic peptides.
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
Spécification
| Laminin alpha-4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P97927, Q16363 | |
| LAMA4 | |
| Recombinant protein fragment of Human LAMA4 (Product #RP-90731) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL3183 | |
| Unconjugated | |
| LAMA4 | |
| CMD1JJ; LAMA3; LAMA4; LAMA4*-1; laminin [a]4; laminin alpha 4 chain; laminin subunit alpha 4; laminin subunit alpha-4; laminin, alpha 4; laminin-14 subunit alpha; Laminin-8 subunit alpha; laminin-9 subunit alpha; RGD1560062 | |
| Mouse | |
| Protein A | |
| RUO | |
| 16775, 309816, 3910 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.