missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin beta 1 Antibody (CL2970) [Alexa Fluor™ 700], Novus Biologicals™
Tous les produits Bio Techne ProduitsDescription
Laminin beta 1 Monoclonal antibody specifically detects Laminin beta 1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigène | Laminin beta 1 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL2970 |
| Conjugué | Alexa Fluor 700 |
| Formule | 50mM Sodium Borate |
| Alias de gène | CLM, cutis laxa with marfanoid phenotype, Laminin B1 chain, laminin subunit beta-1, laminin, beta 1, Laminin-1 subunit beta, Laminin-10 subunit beta, Laminin-12 subunit beta, Laminin-2 subunit beta, Laminin-6 subunit beta, Laminin-8 subunit beta, MGC142015 |
| Espèces hôtes | Mouse |
| Immunogène | This antibody was developed using a recombinant protein derived from P07942, with the exact immunogen sequence remaining proprietary. |
| Méthode de purification | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?