missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAP1 Antibody (RL13) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
213.00€
Spécification
| Antigène | LAP1 |
|---|---|
| Clone | RL13 |
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1:200, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Western Blot |
Description
LAP1 Monoclonal specifically detects LAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Spécification
| LAP1 | |
| 1 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| lamina-associated polypeptide 1B, torsin A interacting protein 1 | |
| TOR1AIP1 | |
| IgG1 | |
| Protein G purified | |
| Detects lamin associated polypeptide (LAP) 1A, 1B & 1C from rat tissues. This does not cross-react with LAP 2.This detects 3 distinct proteins of molecular weights 75, 68 and 55 kDa representing LAP 1A, 1B and 1C, respectively in rat liver nuclear envelope fractions. Immunofluorescence staining of LAP 1 in rat liver with NB120-2737 results in exclusive labeling of the nuclear periphery and exhibits co-localization with lamin staining. |
| RL13 | |
| Western Blot 1:200, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 208263 | |
| Pore complex-lamina fraction isolated from rat liver nuclear envelopes. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit