missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lck Antibody (33D196), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB100-56678AF532
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Lck Monoclonal antibody specifically detects Lck in Human, Mouse samples. It is validated for Western Blot
Spécification
| Lck | |
| Monoclonal | |
| Alexa Fluor 532 | |
| EC 2.7.10, EC 2.7.10.2, Leukocyte C-terminal Src kinase, LSK, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16 | |
| This monoclonal antibody was raised against a synthetic peptide containing amino acid residues of human lck protein. | |
| 0.2 mL | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 3932 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| 33D196 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu