missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LCN1 Monoclonal Antibody (10B10)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA515501
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA5-15501 targets LCN1 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15501 immunogen is purified recombinant fragment of LCN1 expressed in E. Coli.. MA5-15501 detects LCN1 which has a predicted molecular weight of approximately 19.2kDa.
The protein encoded by this gene belongs to the lipocalin family. Lipocalins are a group of extracellular proteins that are able to bind lipophiles by enclosure within their structures to minimize solvent contact. This protein may bind hydrophobic ligands and inhibit cysteine proteinases. It may also play a role in taste reception.
Spécification
| LCN1 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| P31025 | |
| LCN1 | |
| Purified recombinant fragment of LCN1 expressed in E. Coli. | |
| RUO | |
| 3933 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 10B10 | |
| Unconjugated | |
| LCN1 | |
| LCN1; lipocalin 1; lipocalin 1 (tear prealbumin); lipocalin-1; MGC71975; PMFA; protein migrating faster than albumin; Tear lipocalin; Tear prealbumin; Tlc; TP; VEG protein; VEGP; von Ebner gland protein | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu