missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leucyl-cystinyl Aminopeptidase/LNPEP Antibody (E2) [PE], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Leucyl-cystinyl Aminopeptidase/LNPEP Monoclonal specifically detects Leucyl-cystinyl Aminopeptidase/LNPEP in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigène | Leucyl-cystinyl Aminopeptidase/LNPEP |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | E2 |
| Conjugué | PE |
| Formule | PBS |
| Alias de gène | angiotensin IV receptor, AT (4) receptor, CAP, Cystinyl aminopeptidase, EC 3.4.11, EC 3.4.11.3, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, leucyl/cystinyl aminopeptidase, leucyl-cystinyl aminopeptidase, otase, Oxytocinase, Placental leucine aminopeptidase, PLAP, P-LAPinsulin-regulated aminopeptidase, vasopressinase |
| Espèces hôtes | Rabbit |
| Immunogène | Synthetic peptide derived from human Leucyl-cystinyl Aminopeptidase/LNPEP (YFAATQFEPLAAR) |
| Méthode de purification | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?