missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMO2 Recombinant Rabbit Monoclonal Antibody (9H9)
Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA549991
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LMO2 Recombinant Monoclonal Antibody for Western Blot, ELISA
LMO2 encodes a cysteine-rich, two LIM-domain protein that is required for yolk sac erythropoiesis. The LMO2 protein has a central and crucial role in hematopoietic development and is highly conserved. The LMO2 transcription start site is located approximately 25 kb downstream from the 11p13 T-cell translocation cluster (11p13 ttc), where a number T-cell acute lymphoblastic leukemia-specific translocations occur. Alternative splicing results in multiple transcript variants encoding different isoforms.
Spécification
| LMO2 | |
| Recombinant Monoclonal | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P25791 | |
| Lmo2 | |
| A synthesized peptide derived from human LMO2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9H9 | |
| Unconjugated | |
| Lmo2 | |
| cysteine-rich protein TTG-2; LIM domain only 2; LIM domain only 2 (rhombotin-like 1); LIM domain only protein 2; LIM domain only-2; LIM only 2; LMO2; LMO-2; LMO2 protein; Rbtn2; Rbtn-2; RBTNL1; RHOM2; Rhom-2; rhombotin 2; rhombotin-2; rhombotin-like 1; si:dkey-10o6.5; T-cell translocation gene 2; T-cell translocation protein 2; TTG2; wu:fc83a08; zgc:111930; zlmo2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4005 | |
| -20°C or -80°C if preferred | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu