missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMTK2 Monoclonal Antibody (465222)

Rat Monoclonal Antibody
Marque: Invitrogen™ MA561525
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
Spécification
| LMTK2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8IWU2 | |
| LMTK2 | |
| E. coli-derived recombinant human LMTK2, Ala93-Tyr284. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 465222 | |
| Unconjugated | |
| LMTK2 | |
| 2900041G10Rik; A330101P12Rik; aatyk2; Apoptosis-associated tyrosine kinase 2; brain-enriched kinase; BREK; CDK5/p35-regulated kinase; cprk; cyclin-dependent kinase 5/p35-regulated kinase; FLJ46659; hBREK; Kiaa1079; kinase phosphatase inhibitor 2; kinase/phosphatase/inhibitor 2; KPI2; KPI-2; KPI-2 protein; Lemur tyrosine kinase 2; LMKT2; LMR2; Lmtk2; PPP1R100; protein phosphatase 1, regulatory subunit 100; serine/threonine-protein kinase KPI-2; Serine/threonine-protein kinase LMTK2 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 22853 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu