Learn More
LPCAT1 Mouse anti-Human, CoraLite 488, Clone: 8B6E9, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech CL488-66044-100UL
Description
The LPCAT1 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human LPCAT1. This antibody recognizes human, mouse, rat antigen. The LPCAT1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Spécification
| LPCAT1 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Immunofluorescence | |
| 8B6E9 | |
| CoraLite 488 | |
| 100 μL | |
| 79888 | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.