missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-49D Rat anti-Mouse, BUV737, Clone: 400000, BD Biosciences™
Rat Monoclonal Antibody
Marque: BD Biosciences 748496
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Ly-49D Monoclonal antibody specifically detects Ly-49D in Mouse samples. It is validated for Flow Cytometry
Spécification
| Ly-49D | |
| Monoclonal | |
| Brilliant Ultraviolet 737 | |
| Ly49d; Ly-49d; Klra4; Chok | |
| Not Reported | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 400000 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu