missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6B.2 Antibody (7/4) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 13 publications
Marque: Novus Biologicals NBP2-13077-0.025MG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Ly-6B.2 Monoclonal specifically detects Ly-6B.2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Spécification
| Ly-6B.2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Rat | |
| Protein G purified | |
| RUO | |
| recognises Ly6B.2, a polymorphic 40kD antigen expressed by polymorphonuclear cells, but absent on resident tissue macrophages. Strains positive for 7/4:129J; AKR; C57BL/6; C57BL/10; C58; DBA/2; NZB; NZW; SJL; MFI; Swiss (PO) Strains Negative/Weak for 7/4:A2G; A/Sn; ASW; BALB/c; C3H/HEH: CBA.T6T6. | |
| Store at 4C. Do not freeze. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 44016 | |
| Unconjugated | |
| Phosphate buffered saline with 0.09% Sodium Azide | |
| Cultured bone marrow cells | |
| 0.025 mg | |
| Primary | |
| Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu