Learn More
Ly-6B.2 Antibody (7/4) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 13 publications
Marque:  Novus Biologicals NBP2-13077
Code nomenclature Nacres: NA.46
Code produit. 18716593
  • 499.00€ / 125µg
Description
Ly-6B.2 Monoclonal specifically detects Ly-6B.2 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
For Research Use Only