Learn More
LYAG/GAA Antibody, R&D Systems™
Code produit 18620676 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
25 μL
0.1 mL
Conditionnement:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18620676 25 μL 25µL
18074334 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18620676 Fournisseur Novus Biologicals Code fournisseur NBP23814225ul
 Article restreint
Expédition estimée: 01-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
LYAG/GAA Polyclonal specifically detects LYAG/GAA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Spécification
Antigène LYAG/GAA
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Dilution Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formule PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Numéro d’ordre du gène P10253
Alias de gène Acid maltase, Aglucosidase alfa, EC 3.2.1.20, glucosidase, alpha; acid, LYAG, lysosomal alpha-glucosidase
Symboles de gène(s) GAA
Espèces hôtes Rabbit
Immunogène This antibody was developed against a recombinant protein corresponding to amino acids: VGYPFMPPYWGLGFHLCRWGYSSTAITRQVVENMTRAHFPLDVQWNDLDYMDSRRDFTFNKDGFRDFPAMVQELHQGGRRYMMIVDPAISSSGPAGSYRPYDEGLRRGVFITNETGQPLIGKVWPGSTAFPDFTNPTA
Méthode de purification Affinity Purified
Quantité 25 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 2548
Spécificité du test Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Type de produit Antibody
Isotype IgG
Gamme de produits Novus