missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP1-97310AF647
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Polyclonal specifically detects Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Q9UGL1 | |
| KDM5B | |
| A synthetic peptide made to an internal portion of the human JARID1B protein (between residues 800-850) [UniProt Q9UGL1] | |
| 0.1 mL | |
| Cancer, Chromatin Modifiers, Epigenetics | |
| 10765 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu