missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYVE-1 Antibody (223322), PE/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Marque: Novus Biologicals FAB2125PECY7
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LYVE-1 Monoclonal antibody specifically detects LYVE-1 in Mouse samples. It is validated for Flow Cytometry
Spécification
| LYVE-1 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 223322 | |
| Flow Cytometry | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1, UNQ230/PRO263, XLKD1 | |
| BaF/3 mouse pro-B cell line transfected with mouse LYVE-1, Ala24-Thr234, Accession # Q8BHC0 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Endosome Markers, Endothelial Cell Markers, Immunology, Inflammation, Metastasis | |
| 10894 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu