Learn More
MAGEA3 Monoclonal Antibody (OTI24B6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Marque: Origene Technologies TA800832
Description
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita.Spécification
| MAGEA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P43357 | |
| MAGEA3 | |
| Full length human recombit protein of human MAGEA3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI24B6 | |
| Unconjugated | |
| MAGEA3 | |
| CT1.3, HIP8, HYPD, MAGE3, MAGEA6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4102 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.