Learn More
MAP1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21633-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT.Spécification
| MAP1B | |
| Polyclonal | |
| Unconjugated | |
| MAP1B | |
| MAP 1B, MAP1B, MAP5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17755, 4131 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence | |
| 0.49 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P14873, P46821 | |
| Map1b | |
| MAP1B Fusion Protein Ag16255 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |