missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAP7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA556234
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: IAKGALTGGT EVSALPCTTN APGNGKPVGS PHVVTSHQSK VTVESTPDLE KQPNENGVSV Q Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 62%.
The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Spécification
| MAP7 | |
| Polyclonal | |
| Unconjugated | |
| MAP7 | |
| dJ325F22.2 (microtubule-associated protein 7 (EMAP115, E-MAP-115)); EMAP115; E-MAP-115; Ensconsin; epithelial microtubule-associated protein of 115 kDa; MAP7; MAP-7; microtubule associated protein 7; microtubule-associated protein 7; mshi; mste; Mtap7; R75000; ste | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9053 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14244 | |
| MAP7 | |
| Recombinant protein corresponding to Human MAP7. Recombinant protein control fragment (Product #RP-94844). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu