missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mazF Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5144386
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
mazF Polyclonal Antibody for Western Blot, ELISA
Toxic component of a type II toxin-antitoxin (TA) system. MazF is a sequence-specific endoribonuclease that inhibits protein synthesis and induces bacterial stasis. It is very stable, single-strand specific and cleavage is independent of the ribosome. The endoribonuclease activity (a toxin) is inhibited by the labile cognate antitoxin MazE. Toxicity results when the levels of MazE decrease in the cell, leading to mRNA degradation. Both MazE and MazE-MazF bind to the promoter region of the mazE-mazF operon to inhibit their transcription.
Spécification
| mazF | |
| Polyclonal | |
| Unconjugated | |
| mazF | |
| chpA | |
| Rabbit | |
| Protein G | |
| RUO | |
| 75203827 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P0AE71 | |
| mazF | |
| Recombinant Escherichia coli O157:H7 mazF protein (1-111aa ). | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu