missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MDC1 Monoclonal Antibody (P2B11), PE

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA545287
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
0.5 μg/mL of MA5-45287 was sufficient for detection of MDC1 in 10 μg of HeLa cell lysate by ECL immunoblot analysis. Detects approximately 184kDa. This antibody recognizes MDC1 at and around the N-terminus.
MDC1 participates in the early response to DNA damage. It is thought to be a member of the BRCT (BRCA1 C-terminus) super family of nuclear proteins. It contains an N-terminus forkhead-associated (FHA) motif, two C-terminus BRCT motifs and 13 internal repetitions of a 41 amino acid sequence.
Spécification
| MDC1 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Q14676, Q5PSV9, Q7YR40 | |
| MDC1 | |
| GST-tagged recombinant protein corresponding to mouse MDC1 at and around the N-terminus. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Bovine, Chimpanzee | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| P2B11 | |
| PE | |
| MDC1 | |
| 6820401C03; AA413496; DAAP-285E11.7; DKFZp781A0122; homologue to Drosophila photoreceptor protein calphotin; KIAA0170; MDC1; mediation of DNA damage checkpoint 1; mediator of DNA damage checkpoint 1; mediator of DNA damage checkpoint protein 1; mediator of DNA-damage checkpoint 1; MGC166888; mKIAA0170; NFBD1; nuclear factor with BRCT domains 1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 240087, 471959, 511168, 9656 | |
| 4°C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu