missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDMX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 32 publications
Marque: Novus Biologicals NB100-556
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MDMX Polyclonal specifically detects MDMX in Human samples. It is validated for Western Blot, Immunoprecipitation.
Spécification
| MDMX | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZp781B1423, double minute 4 homolog, Double minute 4 protein, double minute 4, human homolog of; p53-binding protein, HDMX, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 4, p53 binding protein, Mdm4, transformed 3T3 cell double minute 4, p53 binding protein (mouse), MDM4-related protein 1, MDMXMGC132766, mouse double minute 4, human homolog of; p53-binding protein, MRP1, p53-binding protein Mdm4, protein Mdm4, Protein Mdmx | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reactivity with mouse MdmX is unlikely owing to divergence of sequence between MdmX and HdmX/MDM4 at the site to which the epitope maps. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Crab-eating macaque and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot, Immunoprecipitation 2 -10 ug/mg lysate | |
| O15151 | |
| MDM4 | |
| The immunogen recognized by this antibody maps to a region between residues 125 and 175 of human ortholog of mouse double minute 4 using the numbering given in Swiss-Prot entry O15151 (GeneID 4194). | |
| 0.1 mL | |
| DNA Repair, Signal Transduction | |
| 4194 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu