missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP3-35641-100ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MED7 Polyclonal antibody specifically detects MED7 in Human,Mouse samples. It is validated for ELISA,Western Blot
Spécification
| MED7 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Activator-recruited cofactor 34 kDa component, Cofactor required for Sp1 transcriptional activation subunit 9, cofactor required for Sp1 transcriptional activation, subunit 9 (33kD), cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa, CRSP33CRSP complex subunit 9, CRSP9hMED7, mediator complex subunit 7ARC34, mediator of RNA polymerase II transcription subunit 7, MGC12284, RNA polymerase transcriptional regulation mediator subunit 7 homolog, Transcriptional coactivator CRSP33 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-233 of human MED7 (NP_004261.1).,, Sequence:, MGEPQQVSALPPPPMQYIKEYTDENIQEGLAPKPPPPIKDSYMMFGNQFQCDDLIIRPLESQGIERLHPMQFDHKKELRKLNMSILINFLDLLDILIRSPGSIKREEKLEDLKLLFVHVHHLINEYRPHQARETLRVMMEVQKRQRLETAERFQKHLERVIEMIQNCLASLPDDLPHSEAGMRVKTEPMDADDSNNCTGQNEHQRENSGHRRDQIIEKDAALCVLIDEMNERP | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 9443 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu