missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12382-1-AP-150UL
This item is not returnable.
View return policy
Description
The proteins belonging to structural maintece of chromosomes (SMC) family play critical roles in chromosome structural changes. SMC proteins are involved in the proper condensation and segregation of mitotic chromosomes. Four human SMC proteins that form two distinct heterodimeric complexes in the cell, the human chromosome-associated protein (hCAP)-C and hCAP-E protein complex (hCAP-C/hCAP-E), and the human SMC1 (hSMC1) and hSMC3 protein complex (hSMC1/hSMC3) have been identified.Specifications
| MEF2A | |
| Polyclonal | |
| Unconjugated | |
| MEF2A | |
| ADCAD1, MEF2, MEF2A, myocyte enhancer factor 2A, RSRFC4, RSRFC9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17258, 309957, 4205 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q02078, Q2MJT0, Q60929 | |
| Mef2a | |
| MEF2A Fusion Protein Ag3049 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction