missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Melan-A Monoclonal Antibody (A103)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA514168
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA5-14168 targets MART-1 in IHC (P) and WB applications and shows reactivity with Human samples. The MA5-14168 immunogen is recombinant MART-1 protein.
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
Spécification
| Melan-A | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| Q16655 | |
| MLANA | |
| Recombinant MART-1 protein. | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| A103 | |
| Unconjugated | |
| MLANA | |
| A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
| Mouse | |
| Protein G | |
| RUO | |
| 2315 | |
| 4°C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu