missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MERS Coronavirus Spike Protein S1 Monoclonal Antibody (3871)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA533278
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Does not cross react with the SARS Coronavirus.
The spike (S) glycoprotein of coronaviruses contains protrusions that will only bind to certain receptors on the host cell: they are essential for both host specificity and viral infectivity. The term 'peplomer' is typically used to refer to a grouping of heterologous proteins on the virus surface that function together. The spike (S) glycoprotein of coronaviruses is known to be essential in the binding of the virus to the host cell at the advent of the infection process. Most notable is severe acute respiratory syndrome (SARS). The severe acute respiratory syndrome-coronavirus (SARS-CoV) spike (S) glycoprotein alone can mediate the membrane fusion required for virus entry and cell fusion. It is also a major immunogen and a target for entry inhibitors. The SARS-CoV spike (S) protein is composed of two subunits; the S1 subunit contains a receptor-binding domain that engages with the host cell receptor angiotensin-converting enzyme 2 and the S2 subunit mediates fusion between the viral and host cell membranes. The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity, during infection with SARS-CoV.
Spécification
| MERS Coronavirus Spike Protein S1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 3871 | |
| Unconjugated | |
| MERS Coronavirus S protein; Spike glycoprotein | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu