missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I (H-2Db) Monoclonal Antibody (28-14-8), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
160.00€ - 311.00€
Spécification
| Antigène | MHC Class I (H-2Db) |
|---|---|
| Clone | 28-14-8 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| MHC Class I (H-2Db) | |
| 0.1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2DB; H2-DC1-beta transplantation antigen protein; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein | |
| H2-D1 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| H2-D1 |
| 28-14-8 | |
| Flow Cytometry | |
| NovaFluor Blue 660-120S | |
| Mouse | |
| Mouse | |
| P01895, P01896, P01897, P01899, P01900, P14426, P14427 | |
| 14964 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title