missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I (H-2Kb) Monoclonal Antibody (AF6-88.5.5.3), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
191.00€ - 359.00€
Spécification
| Antigène | MHC Class I (H-2Kb) |
|---|---|
| Clone | AF6-88.5.5.3 |
| Concentration | 0.1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Spécification
| MHC Class I (H-2Kb) | |
| 0.1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2-K1; histocompatibility 2, K1, K region; K-f; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen | |
| H2-K1 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
| AF6-88.5.5.3 | |
| Flow Cytometry | |
| NovaFluor Red 700 | |
| Mouse | |
| Mouse | |
| P01901, P03991, P04223, P14428 | |
| 14972 | |
| IgG2a κ | |
| Affinity chromatography | |
| H2-K1 |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit