missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Antibody (P7/7) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-09015-0.025mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MHC Class II Monoclonal antibody specifically detects MHC Class II in Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Spécification
| MHC Class II | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| P7/7 | |
| Unconjugated | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| This antibody was raised by immunising DA rats with PVG.R8 rat spleen cells. Splenocytes from immunised rats were then isolated, and fused to the myeloma line NSO/u to generate stable hybridomas. | |
| 0.025 mg | |
| Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 3108 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu