missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tampon d’étalonnage au millième pH 4,01, Hanna Instruments™
Click to view available options
Quantité:
500 mL
Spécification
Couleur | Incolore |
pH | 4.01 |
Forme physique | Liquide |
Quantité | 500 mL |
Type de solution | Buffer |
Formule | Aqueous Buffer Solution |
Odeur | Odorless |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'