missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIS12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA540125
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Recommended positive controls: HeLa and HepG2 cells. Predicted reactivity: Rhesus Monkey (97%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This protein is part of the MIS12 complex which is required for normal chromosome alignment and segregation and for kinetochore formation during mitosis.
Spécification
| MIS12 | |
| Polyclonal | |
| Unconjugated | |
| MIS12 | |
| 2510025F08Rik; Ac1585; AU044651; hMis12; homolog of yeast Mis12; KNTC2AP; Liver regeneration-related protein LRRG112; Mis12; MIS12 homolog; MIS12 homolog (yeast); MIS12 kinetochore complex component; MIS12, kinetochore complex component; MIS12, MIND kinetochore complex component, homolog; Misc12; MTW1; Protein MIS12 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79003 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.99 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9H081 | |
| MIS12 | |
| Recombinant protein encompassing a sequence within the center region of human MIS12. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu