Learn More
MMR/CD206/Mannose Receptor Antibody (CL0387) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-52927AF488
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MMR/CD206/Mannose Receptor Monoclonal antibody specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spécification
| MMR/CD206/Mannose Receptor | |
| Monoclonal | |
| Alexa Fluor 488 | |
| CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen | |
| This antibody was developed using a recombinant protein derived from P22897, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Immunology, Innate Immunity, Signal Transduction | |
| 4360 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0387 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |