missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse Albumin Polyclonal Antibody
Goat Polyclonal Antibody
Marque: Invitrogen™ PA130899
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PA1-30899 detects Mouse Albumin from mouse samples.
Serum albumin, a globular unglycosylated protein of 65 kDa, is the main protein of plasma, has a good binding capacity for water, fatty acids, hormones, bilirubin and drugs. Its main function is the regulation of the colloidal osmotic pressure of blood. Albumin is synthesized in the liver as preproalbumin, which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to give the secreted albumin.
Spécification
| Mouse Albumin | |
| Polyclonal | |
| Unconjugated | |
| Alb | |
| Alb; Alb1; Alb-1; albumin; albumin 1; serum albumin | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 11657 | |
| 4°C | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| P07724 | |
| Alb | |
| Mouse serum albumin in Freund's adjuvant. | |
| 1 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu